missing translation for 'onlineSavingsMsg'
Learn More

Benztropine Mesylate, MP Biomedicals™

Product Code. 11472982 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11472982 100 mg 100mg
11462982 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11472982 Supplier MP Biomedicals Supplier No. 0215362680
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 132-17-2
Molecular Formula C22H29NO4S
Molecular Weight (g/mol) 403.54
MDL Number MFCD00074784
InChI Key CPFJLLXFNPCTDW-STYNFMPRSA-N
Synonym benztropine mesylate, cogentin, benztropine mesilate, benztropine methanesulfonate, unii-wmj8tl7510, benztropine methylsulfonate, benzatropine mesilate, benzatropine mesylate, benzotropine mesylate, cobrentin methanesulfonate
PubChem CID 3246155
SMILES CS(O)(=O)=O.CN1[C@@H]2CC[C@@H]1CC(C2)OC(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 100 mg
IUPAC Name (1R,5S)-3-benzhydryloxy-8-methyl-8-azabicyclo[3.2.1]octane;methanesulfonic acid
Formula Weight 403.5
Physical Form Solid
Chemical Name or Material Benztropine Mesylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.