missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 3-oxopiperazine-1-carboxylate, 97%, Thermo Scientific™

Product Code. 10265524
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10265524 5 g 5g
10678945 10 g 10g
10613645 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10265524 Supplier Thermo Scientific Maybridge Supplier No. KM07871DE
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 78818-15-2
Molecular Formula C12H14N2O3
Molecular Weight (g/mol) 234.255
InChI Key BAHFPJFBMJTOPU-UHFFFAOYSA-N
Synonym 4-benzyloxycarbonylpiperazin-2-one, 1-cbz-3-oxopiperazine, 4-benzyloxycarbonyl-2-piperazinone, 4-cbz-piperazinone, 1-z-3-oxopiperazine, 4-cbz-piperazin-2-one, 1-piperazinecarboxylic acid, 3-oxo-, phenylmethyl ester, 3-oxopiperazine-1-carboxylic acid benzyl ester, benzyl 3-oxotetrahydro-1 2h-pyrazinecarboxylate, phenylmethyl 3-oxopiperazinecarboxylate
PubChem CID 736777
IUPAC Name benzyl 3-oxopiperazine-1-carboxylate
SMILES C1CN(CC(=O)N1)C(=O)OCC2=CC=CC=C2

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 234.25
Percent Purity 97%
Chemical Name or Material Benzyl 3-oxopiperazine-1-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.