missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-bromopiperidine-1-carboxylate, 96%

Product Code. 10133404 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10133404 1 g Glass bottle 1g
10193444 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10133404 Supplier Thermo Scientific Acros Supplier No. 427030010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 166953-64-6 | C13H16BrNO2 | 298.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 166953-64-6
Molecular Formula C13H16BrNO2
Molecular Weight (g/mol) 298.18
MDL Number MFCD02681990
InChI Key BFWKSPOSMPKTSV-UHFFFAOYSA-N
Synonym benzyl 4-bromotetrahydro-1 2h-pyridinecarboxylate, n-cbz-4-bromo-piperidine, 4-bromo-n-z-piperidine, 4-bromopiperidine, n-cbz protected, 1-cbz-4-bromopiperidine, benzyl4-bromopiperidine-1-carboxylate, benzyl 4-bromo-1-piperidinecarboxylate, pubchem16157, buttpark 9350-52, 4-bromo-n-cbz piperidine
PubChem CID 2776275
IUPAC Name benzyl 4-bromopiperidine-1-carboxylate
SMILES C1CN(CCC1Br)C(=O)OCC2=CC=CC=C2

Specifications

CAS Min % 95.0
CAS Max % 100.0
Density 1.3740g/mL
Boiling Point 144°C (0.3 mmHg)
Flash Point >110°C
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.374
Formula Weight 298.18
Percent Purity 96%
Chemical Name or Material Benzyl 4-bromopiperidine-1-carboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Benzyl 4-bromopiperidine-1-carboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.