missing translation for 'onlineSavingsMsg'
Learn More

Benzyl alcohol, Solstice

Product Code. 15607170
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15607170 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15607170 Supplier Honeywell-Chromasolv Supplier No. 807081L
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

CHROMASOLV™, GC-Headspace tested,≥99.9% (GC)

Solstice Chromasolv™ GC-headspace solvents:

  • developed particularly for the identification and quantification of residual solvents in pharmaceuticals using GC-headspace methods described in monographs from the United States Pharmacopeia (USP) or the European Pharmacopoeia (Ph. Eur.) and following the International Conference on Harmonization of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH) guidelines
  • very pure solvents with extremely low concentrations of the defined residual solvents
  • state-of-the art production and application tests
TRUSTED_SUSTAINABILITY

Specifications

CAS 100-51-6
Density 1.045g/mL at 25°C
Flash Point 96°C (204.8°F)
Ignition Point 817°F
Molecular Formula C7H8O
Linear Formula C6H5CH2OH
MDL Number MFCD00004599,MFCD03792087
Quantity 1 L
UN Number UN3334
Beilstein 878307
Synonym benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
SMILES OCC1=CC=CC=C1
IUPAC Name phenylmethanol
Molecular Weight (g/mol) 108.14
PubChem CID 244
Vapor Density 3.7 (vs air)
ChEBI CHEBI:17987
Formula Weight 108.14g/mol
Vapor Pressure 13.3 mmHg (100°C), 3.75 mmHg (77°C)
Percent Purity ≥99.9% (GC)
Grade CHROMASOLV
Chemical Name or Material Benzyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • Benzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302+H332-Harmful if swallowed or if inhaled
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.