missing translation for 'onlineSavingsMsg'
Learn More

Benzyl alcohol, MP Biomedicals™

Product Code. 11446281 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11446281 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11446281 Supplier MP Biomedicals Supplier No. 0219388290
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Benzyl alcohol ACS reagent grade, Clear Liquid

  • It may be used for the benzylation of toluene.
  • Benzyl alcohol (BnOH) is a monoaromatic primary alcohol widely used as a solvent and an intermediate in cosmetic formulations, pharmaceutical and flavor/fragrance industries.
  • It shows bacteriostatic property.
  • Commercial method for its synthesis is by the alkaline hydrolysis of benzyl chloride.
  • Conformational studies by ab initio calculations shows gauche cis conformation around the alcoholic group.

Specifications

CAS 100-51-6
Melting Point °C to 15.3°C (literature)
Color ≤20 (APHA)
Density 1.0442g/cm3 at 22.5°C (Lit.)
Boiling Point 205.4°C (at 1013 hPa (Literature))
Flash Point 204.8°F/96°C (Lit.)
Molecular Formula C7H8O
Refractive Index 0.03 hPa at 20°C ; 13.3 mmHg ( 100°C), 3.75 mmHg ( 77°C) (Lit.)
MDL Number MFCD00004599,MFCD03792087
Quantity 500 mL
Beilstein 878307
Synonym Benzene Carbinol, Benzenemethanol, Benzoyl Alcohol, Hydroxytoluene
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
SMILES OCC1=CC=CC=C1
IUPAC Name phenylmethanol
Molecular Weight (g/mol) 108.14
Vapor Density 3.7 (vs air) (Lit.)
Vapor Pressure 0.03 hPa at 20°C ; 13.3 mmHg ( 100°C), 3.75 mmHg ( 77°C) (Lit.)
Grade ACS Reagent
Physical Form Liquid
Assay 0.999
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.