missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alcohol, Multi-Compendial, N.F., J.T. Baker™

Product Code. 15528514 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Unit Size:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15528514 4 L Narrow Mouth Amber Glass Bottle 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15528514 Supplier Avantor J.T.Baker Supplier No. 903914
 Restricted Item
Estimated Shipment: 10-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 100-51-6
Flash Point 93.00°C
Packaging Narrow Mouth Amber Glass Bottle
Molecular Formula C7H8O
MDL Number MFCD00004599,MFCD03792087
Quantity 4 L
Specific Gravity 1L = 1.05kg
Synonym benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
SMILES OCC1=CC=CC=C1
IUPAC Name phenylmethanol
Molecular Weight (g/mol) 108.14
PubChem CID 244
ChEBI CHEBI:17987
Formula Weight 108.14
Grade Multi-Compendial/NF
Chemical Name or Material Benzyl Alcohol
Show More Show Less
compliance-icons
Product Identifier
  • Benzyl Alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Benzyl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.