missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Alcohol, puriss., Honeywell

Click to view available options
Quantity:
1 L
2.5 L
30 kg
Unit Size:
1 L
2.5 L
30kg
Tekniske data
Tekniske data
| Density | 1.045g/mL at 25°C |
| Flash Point | 96°C (204.8°F) |
| Ignition Residue | ≤0.005% (as SO4) |
| Ignition Point | 817°F |
| Linear Formula | C6H5CH2OH |
| Quantity | 30 kg |
| UN Number | 3334 |
| Beilstein | 878307 |
| Vapor Density | 3.7 (vs air) |
| Formula Weight | 108.14g/mol |
| Vis mere |
Safety and Handling
Product Identifier
- Benzyl alcohol
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H302 + H332-Harmful if swallowed or if inhaled.
- H319-Causes serious eye irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction