missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alternariol, TRC

Product Code. 30346625
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30346625 100 mg 100mg
30348387 25 mg 25mg
30342053 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30346625 Supplier Toronto Research Chemicals Supplier No. TRCB209975100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl Alternariol
Analyte or Component Names Benzyl Alternariol
Shipping Condition Room Temperature
Molecular Formula C21 H16 O5
Synonym 9-(Benzyloxy)-3,7-dihydroxy-1-methyl-6H-benzo[c]chromen-6-one
InChI Formula InChI=1S/C21H16O5/c1-12-7-14(22)8-18-19(12)16-9-15(10-17(23)20(16)21(24)26-18)25-11-13-5-3-2-4-6-13/h2-10,22-23H,11H2,1H3
Solution Type Neat
SMILES Cc1cc(O)cc2OC(=O)c3c(O)cc(OCc4ccccc4)cc3c12
IUPAC Name 3,7-dihydroxy-1-methyl-9-phenylmethoxybenzo[c]chromen-6-one
Industry Type Pharmaceutical
Molecular Weight (g/mol) 348.349
Quantity 100 mg
Formula Weight 348.1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.