missing translation for 'onlineSavingsMsg'
Learn More

Benzyl azide, 94%

Product Code. 11382878
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11382878 10 g 10g
11392878 2 g 2g
11372878 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11382878 Supplier Thermo Scientific Alfa Aesar Supplier No. 032635.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 622-79-7 | C7H7N3 | 133.154 g/mol

Benzyl azide is used in the synthesis of 1,2,3-triazole derivatives. It serves as a reagent used to introduce a PhCH2N group into a molecule.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-79-7
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.154
MDL Number MFCD00013836
InChI Key UDLLFLQFQMACJB-UHFFFAOYSA-N
Synonym azidomethyl benzene, benzyl azide, benzylazide, benzene, azidomethyl, alpha-azidotoluene, toluene, .alpha.-azido, unii-hfd57z7j9j, .alpha.-azidotoluene, ccris 8029
PubChem CID 12152
IUPAC Name azidomethylbenzene
SMILES C1=CC=C(C=C1)CN=[N+]=[N-]

Specifications

Density 1.0655 g/mL
Boiling Point 82°C to 85°C (16 mmHg)
Flash Point 43°C (110°F)
Assay Percent Range 94%
Refractive Index 1.5341
Quantity 10 g
Solubility Information Insoluble in waterMiscible with ethanol and diethyl ether. Immiscible with water.
Formula Weight 133.15
Physical Form Liquid
Chemical Name or Material Benzyl Azide
compliance-icons
Product Identifier
  • Benzyl azide
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Benzyl azide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.