missing translation for 'onlineSavingsMsg'
Learn More

Benzyl benzoate, 99+%

Product Code. 11372076
Change view
Click to view available options
Quantity:
250 g
1000 g
2500 g
Unit Size:
1kg
2.5kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372076 250 g 250g
11382076 1000 g 1kg
11392076 2500 g 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatorischer Hinweis: Dieses Produkt ist ein isoliertes Zwischenprodukt, das gemäß der Verordnung (EG) Nr. 1907/2006 (REACH) transportiert wird. Alle Kunden müssen vor der Lieferung eine Erklärung ausfüllen.
Artikelnummer. 11372076 Lieferant Thermo Scientific Alfa Aesar Lieferanten-Nr. L03258.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Dieses Produkt ist ein isoliertes Zwischenprodukt, das gemäß der Verordnung (EG) Nr. 1907/2006 (REACH) transportiert wird. Alle Kunden müssen vor der Lieferung eine Erklärung ausfüllen.

CAS: 120-51-4 | C14H12O2 | 212.25 g/mol

Benzyl benzoate is used as repellent for chiggers, ticks, and mosquitoes. It is an effective and inexpensive topical treatment for human scabies. It has vasodilating and spasmolytic effects and is present in many asthma and whooping cough drugs. It is also a dye carrier, solvent for cellulose derivatives, plasticizer, and fixative in the perfume industry

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Miscible with ethanol, alcohol, chloroform, ether, oils.

Notes
Incompatible with oxidizing agents. Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 120-51-4
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00003075
InChI Key SESFRYSPDFLNCH-UHFFFAOYSA-N
Synonym ascabiol, benylate, novoscabin, benzoic acid, phenylmethyl ester, scabitox, scobenol, ascabin, benzyl phenylformate, benzoic acid, benzyl ester, phenylmethyl benzoate
PubChem CID 2345
ChEBI CHEBI:41237
IUPAC Name benzyl benzoate
SMILES O=C(OCC1=CC=CC=C1)C1=CC=CC=C1

Spezifikation

Melting Point 18°C to 20°C
Density 1.112
Boiling Point 323°C to 324°C
Flash Point 147°C (296°F)
Linear Formula C6H5CO2CH2C6H5
Refractive Index 1.568
Quantity 250 g
UN Number UN3082
Beilstein 2049280
Merck Index 14,1127
Solubility Information Miscible with ethanol,alcohol,chloroform,ether,oils.
Formula Weight 212.25
Percent Purity ≥99%
Chemical Name or Material Benzyl benzoate
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Produktidentifikator
  • Benzyl benzoate
Signalwort
  • Warning
Gefahrenkategorie
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
Gefahrenhinweis
  • H302-Harmful if swallowed.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Sicherheitshinweis
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Ergänzende Informationen
  • MIXTURE LIST-Contain: Benzyl benzoate

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.