missing translation for 'onlineSavingsMsg'
Learn More

Benzyl benzoate, 99+%

Product Code. 10607744 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
250mL
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10607744 5 L Plastic drum 5 L
10654752 250 mL Glass bottle 250mL
10396210 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10607744 Supplier Thermo Scientific Acros Supplier No. 105860051
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 120-51-4 | C14H12O2 | 212.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-51-4
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00003075
InChI Key SESFRYSPDFLNCH-UHFFFAOYSA-N
Synonym ascabiol, benylate, novoscabin, benzoic acid, phenylmethyl ester, scabitox, scobenol, ascabin, benzyl phenylformate, benzoic acid, benzyl ester, phenylmethyl benzoate
PubChem CID 2345
ChEBI CHEBI:41237
IUPAC Name benzyl benzoate
SMILES O=C(OCC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Acidity 0.3mg KOH/g max.
CAS Min % 99.0
CAS Max % 100.0
Melting Point 18.0°C to 20.0°C
Color Colorless
Density 1.1180g/mL
Boiling Point 323.0°C
Flash Point 147°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic drum
Linear Formula C6H5CO2CH2C6H5
Refractive Index 1.5670 to 1.5700
Quantity 5 L
Beilstein 09, 121
Merck Index 15, 1130
Specific Gravity 1.118
Solubility Information Solubility in water: 15.3mg/l (20°C). Other solubilities: soluble in alcohol,chloroform and ether,insoluble in glycerol
Viscosity 10.9 mPa.s (25°C)
Formula Weight 212.25
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Benzyl benzoate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Benzyl benzoate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.