missing translation for 'onlineSavingsMsg'
Learn More

Benzyl benzoate, MedChemExpress

Product Code. 16438526
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16438526 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16438526 Supplier Medchem Express Supplier No. HYB0935
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Benzyl benzoate (Benzoic acid benzyl ester) is a fragrance ingredient in cosmetic products. Benzyl benzoate can be used for the research of Scabies and Demodex-associated inflammatory skin conditions.

Benzyl benzoate (Benzoic acid benzyl ester) is a fragrance ingredient in cosmetic products. Benzyl benzoate can be used for the research of Scabies and Demodex-associated inflammatory skin conditions.

Specifications

Chemical Name or Material Benzyl benzoate
Physical Form Liquid
CAS 120-51-4
Color Colorless
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C14H12O2
For Use With (Application) Neuroscience-Neuromodulation
Synonym Benzoic acid benzyl ester
Solubility Information DMSO : ≥ 50 mg/mL (235.58 mM)
SMILES O=C(OCC1=CC=CC=C1)C2=CC=CC=C2
Molecular Weight (g/mol) 212.24
Formula Weight 212.24
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.