missing translation for 'onlineSavingsMsg'
Learn More

Benzyl chloride, 99%, pure, stabilized

Product Code. 15179283 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
250g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15179283 10 kg Plastic drum 10kg
10592052 5 g Glass Bottle 5g
10720482 250 g Glass Bottle 250g
10065740 1 kg Glass Bottle 1kg
10167323 2.5 kg Glass Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15179283 Supplier Thermo Scientific Acros Supplier No. 405090100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 100-44-7 | C7H7Cl | 126.583 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-44-7 , 75-56-9
MDL Number MFCD00000889
InChI Key KCXMKQUNVWSEMD-UHFFFAOYSA-N
Synonym benzyl chloride, chloromethyl benzene, benzylchloride, alpha-chlorotoluene, benzylchlorid, chlorophenylmethane, benzene, chloromethyl, chlorure de benzyle, tolyl chloride, alpha-chlortoluol
PubChem CID 7503
ChEBI CHEBI:615597
IUPAC Name chloromethylbenzene
SMILES C1=CC=C(C=C1)CCl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -39.0°C
Color Colorless to Yellow
Density 1.1000g/mL
Boiling Point 179.0°C
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic drum
Molecular Formula C7H7Cl
Linear Formula C6H5CH2Cl
Refractive Index 1.5370 to 1.5390
Quantity 10 kg
Beilstein 05,292
Merck Index 15,1132
Specific Gravity 1.1
Solubility Information Solubility in water: 0.3g/L (20°C). Other solubilities: miscible with ethanol,ethyl ether,chloroform,,acetone,slightly soluble in carbon tetrachloride
Stabilizer 0.25% propylene oxide
Molecular Weight (g/mol) 126.59
Viscosity 1.280 mPa.s (20°C)
Formula Weight 126.59
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material Benzyl chloride, Stabilized, Pure
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Benzyl chloride, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Germ cell mutagenicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H290-May be corrosive to metals.
  • H318-Causes serious eye damage.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H315-Causes skin irritation.
  • H373-May cause damage to organs.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.