missing translation for 'onlineSavingsMsg'
Learn More

Benzyl chloride, 99%, stab.

Product Code. 11464094
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464094 500 g 500g
11474094 2500 g 2500g
11484094 10,000 g 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11464094 Supplier Thermo Scientific Alfa Aesar Supplier No. A12481.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 100-44-7 | C7H7Cl | 126.583 g/mol

Benzyl Chloride is used as an intermediate for manufacturing of benzyl alcohol, benzoic acid and benzyl cyanide. It is also used to prepare benzyl esters, which are used as plasticizer, flavorants and perfumes. It acts as an intermediate in the preparation of phenylacetic acid. In organic synthesis, it is used as an alkylating agent and utilized for the introduction of the benzyl protecting group for alcohols. Further, it is involved in the preparation of quaternary ammonium salts by reaction with tertiary amines. In addition to this, it is used to prepare Grignard reagent by reacting with metallic magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl Chloride is used as an intermediate for manufacturing of benzyl alcohol, benzoic acid and benzyl cyanide. It is also used to prepare benzyl esters, which are used as plasticizer, flavorants and perfumes. It acts as an intermediate in the preparation of phenylacetic acid. In organic synthesis, it is used as an alkylating agent and utilized for the introduction of the benzyl protecting group for alcohols. Further, it is involved in the preparation of quaternary ammonium salts by reaction with tertiary amines. In addition to this, it is used to prepare Grignard reagent by reacting with metallic magnesium.

Solubility
Miscible with alcohol, ether. Insoluble in waterMiscible with alcohol, ether and chloroform.

Notes
Light and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with common metals, lead,oxidizing agents, iron, iron salts, brass and aluminum
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-44-7
Molecular Formula C7H7Cl
Molecular Weight (g/mol) 126.583
MDL Number MFCD00000889
InChI Key KCXMKQUNVWSEMD-UHFFFAOYSA-N
Synonym benzyl chloride, chloromethyl benzene, benzylchloride, alpha-chlorotoluene, benzylchlorid, chlorophenylmethane, benzene, chloromethyl, chlorure de benzyle, tolyl chloride, alpha-chlortoluol
PubChem CID 7503
ChEBI CHEBI:615597
IUPAC Name chloromethylbenzene
SMILES C1=CC=C(C=C1)CCl

Specifications

Melting Point - 43°C
Density 1.099
Boiling Point 178°C to 180°C
Flash Point 73°C (163°F)
Assay Percent Range 99%, Stabilized
Refractive Index 1.538
Quantity 500 g
UN Number UN1738
Beilstein 471308
Merck Index 14,1129
Solubility Information Miscible with alcohol,ether; Insoluble in waterMiscible with alcohol,ether and chloroform.
Formula Weight 126.59
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Benzyl Chloride, stabilized
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Benzyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Germ cell mutagenicity Category 1B
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Benzyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.