missing translation for 'onlineSavingsMsg'
Learn More

Benzyl salicylate, 99%

Product Code. 11463257
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463257 500 g 500g
11453257 100 g 100g
11473257 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463257 Supplier Thermo Scientific Alfa Aesar Supplier No. A16184.36
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 118-58-1 | C14H12O3 | 228.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-58-1
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.247
MDL Number MFCD00020034
InChI Key ZCTQGTTXIYCGGC-UHFFFAOYSA-N
Synonym benzyl salicylate, benzyl o-hydroxybenzoate, salicylic acid, benzyl ester, benzoic acid, 2-hydroxy-, phenylmethyl ester, phenylmethyl 2-hydroxybenzoate, salicylic acid benzyl ester, salicyclic acid, benzyl ester, salicylsaeurebenzylester, unii-wao5mnk9tu, salicyclic acid benzyl ester
PubChem CID 8363
IUPAC Name benzyl 2-hydroxybenzoate
SMILES C1=CC=C(C=C1)COC(=O)C2=CC=CC=C2O

Specifications

Melting Point 18°C to 20°C
Density 1.17
Boiling Point 190°C (14 mmHg)
Flash Point 137°C (278°F)
Refractive Index 1.5804
Quantity 500 g
Beilstein 2115365
Merck Index 14,1144
Formula Weight 228.25
Percent Purity 99%
Chemical Name or Material Benzyl salicylate
Show More Show Less
compliance-icons
Product Identifier
  • Benzyl salicylate
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Benzyl salicylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.