missing translation for 'onlineSavingsMsg'
Learn More

Benzylamine, 98+%, (Acidimetric), Fisher Chemical

Product Code. 10427250 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10427250 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10427250 Supplier Fisher Chemical Supplier No. B/2600/PB05
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 100-46-9
Melting Point -30°C
pH 11.6
Boiling Point 182°C
Packaging Plastic Coated Glass bottle
Molecular Formula C7H9N
MDL Number 8106
Quantity 100 mL
Vapor Density 3.7
Formula Weight 107.15g/mol
Vapor Pressure 0.6mbar at 20°C
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.