missing translation for 'onlineSavingsMsg'
Learn More

Benzylamine hydrochloride, 99%

Product Code. 10694712 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10694712 5 g Glass bottle 5g
10173830 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10694712 Supplier Thermo Scientific Acros Supplier No. 200540050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3287-99-8 | C7H9N·ClH | 143.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3287-99-8
Molecular Formula C7H9N·ClH
Molecular Weight (g/mol) 143.62
MDL Number MFCD00012852
InChI Key XKXHCNPAFAXVRZ-UHFFFAOYSA-N
Synonym benzylamine hydrochloride, phenylmethanamine hydrochloride, benzenemethanamine, hydrochloride, benzylammonium chloride, benzylamine, hydrochloride, usaf el-82, benzenemethanamine, hydrochloride 1:1, benzylamine hcl, benzylaminehydrochloride, benzyl amine hcl salt
PubChem CID 2724127
IUPAC Name phenylmethanamine;hydrochloride
SMILES C1=CC=C(C=C1)CN.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 259°C to 264°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H5CH2NH2·HCl
Quantity 5 g
Beilstein 12, 1017
Merck Index 14, 1125
Solubility Information Solubility in water: soluble
Formula Weight 143.62
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Benzylamine hydrochloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Benzylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.