missing translation for 'onlineSavingsMsg'
Learn More

Benzyldecyldimethylammonium Chloride, TRC

Product Code. 30134533
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134533 10 g 10g
30003817 1 g 1g
18013830 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134533 Supplier Toronto Research Chemicals Supplier No. TRCB23455010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 965-32-2
Molecular Formula C19 H34 N . Cl
Molecular Weight (g/mol) 311.93
Synonym Benzenemethanaminium, N-decyl-N,N-dimethyl-, chloride (1:1), Ammonium, benzyldecyldimethyl-, chloride (8CI), Benzenemethanaminium, N-decyl-N,N-dimethyl-, chloride (9CI), Benzyldecyldimethylammonium chloride (6CI,7CI), Benzyldimethyldecylammonium chloride, Capryldimethylbenzylammonium chloride, Decylbenzyldimethylammonium chloride, Decyldimethylbenzylammonium chloride, Dimethylbenzyldecylammonium chloride, N-Benzyldimethyldecylammonium chloride, N-Decyl-N,N-dimethylbenzenemethanaminium chloride, Querton 10B-Cl
IUPAC Name benzyl-decyl-dimethylazanium;chloride
SMILES [Cl-].CCCCCCCCCC[N+](C)(C)Cc1ccccc1

Specifications

Analyte or Component Names Benzyldecyldimethylammonium Chloride
Shipping Condition Room Temperature
InChI Formula InChI=1S/C19H34N.ClH/c1-4-5-6-7-8-9-10-14-17-20(2,3)18-19-15-12-11-13-16-19;/h11-13,15-16H,4-10,14,17-18H2,1-3H3;1H/q+1;/p-1
Solution Type Neat
Industry Type Pharmaceutical
Quantity 10 g
Formula Weight 311.238
Percent Purity >95%
Chemical Name or Material Benzyldecyldimethylammonium Chloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.