missing translation for 'onlineSavingsMsg'
Learn More

Benzyldimethyltetradecylammonium Chloride Dihydrate, TRC

Product Code. 30012808
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012808 100 mg 100mg
30005618 50 mg 50mg
30005283 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012808 Supplier Toronto Research Chemicals Supplier No. TRCB535485100MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147228-81-7
Molecular Formula C23 H42 N . Cl . 2 H2 O
Molecular Weight (g/mol) 404.07
Synonym N,N-Dimethyl-N-tetradecylbenzenemethanaminium Chloride Hydrate (1:1:2)
IUPAC Name benzyl-dimethyl-tetradecylazanium;chloride;dihydrate
SMILES O.O.[Cl-].CCCCCCCCCCCCCC[N+](C)(C)Cc1ccccc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C23H42N.ClH.2H2O/c1-4-5-6-7-8-9-10-11-12-13-14-18-21-24(2,3)22-23-19-16-15-17-20-23;;;/h15-17,19-20H,4-14,18,21-22H2,1-3H3;1H;2*1H2/q+1;;;/p-1
Formula Weight 403.322
Chemical Name or Material Benzyldimethyltetradecylammonium Chloride Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.