missing translation for 'onlineSavingsMsg'
Learn More

Benzylideneacetone, TRC

Product Code. 30136256
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136256 10 g 10g
17901955 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136256 Supplier Toronto Research Chemicals Supplier No. TRCB27997510G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-57-6
Molecular Formula C10 H10 O
Molecular Weight (g/mol) 146.19
Synonym 2-Butenone, 4-phenyl- (2CI), Ketone, methyl styryl (7CI), 4-Phenyl-3-buten-2-one, 1-Phenyl-1-buten-3-one, 2-Phenylethenyl methyl ketone, 2-Phenylvinyl methyl ketone, 4-Phenyl-3-butene-2-one, 4-Phenylbutenone, Acetocinnamone, Benzalacetone, Benzylideneacetone, Methyl 2-phenylvinyl ketone, Methyl phenylvinyl ketone, Methyl styryl ketone, Methyl β-styryl ketone, NSC 5605, Styryl methyl ketone
IUPAC Name 4-phenylbut-3-en-2-one
SMILES CC(=O)C=Cc1ccccc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C10H10O/c1-9(11)7-8-10-5-3-2-4-6-10/h2-8H,1H3
Formula Weight 146.0732
Chemical Name or Material Benzylideneacetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.