missing translation for 'onlineSavingsMsg'
Learn More

Benzyltriphenylphosphonium chloride, 99%

Product Code. 11497916
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packungsgröße
11497916 100 g 100g
11487916 25 g 25g
11407926 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11497916 Lieferant Thermo Scientific Alfa Aesar Lieferanten-Nr. A14034.22
 Restricted Item
In Stock
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 1100-88-5 | C25H22ClP | 388.87 g/mol

Benzyltriphenylphosphonium chloride is used as the accelerator of the Bisphenol AF curing system for fluoroelastomer. Also used as phase-transfer catalysts in the synthesis of certain organic compounds. It is used as a reactant for the synthesis of chemical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyltriphenylphosphonium chloride is used as the accelerator of the Bisphenol AF curing system for fluoroelastomer. Also used as phase-transfer catalysts in the synthesis of certain organic compounds. It is used as a reactant for the synthesis of chemical compounds.

Solubility
Soluble in water and alcohol. Insoluble in acetone.

Notes
Hygroscopic. Store under inert gas. Store away from strong oxidizing agents, water/ moisture..
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 1100-88-5
Molecular Formula C25H22ClP
Molecular Weight (g/mol) 388.87
MDL Number MFCD00011913
InChI Key USFRYJRPHFMVBZ-UHFFFAOYSA-M
Synonym benzyltriphenylphosphonium chloride, triphenylbenzylphosphonium chloride, phosphonium, triphenyl phenylmethyl-, chloride, benzyltriphenylphosphanium chloride, benzyl triphenylphosphonium chloride, benzyl-triphenyl-phosphonium chloride, phosphonium, triphenyl phenylmethyl-, chloride 1:1, triphenylbenzylphosphine, chloride, benzyl triphenyl phosphoniumchlorid
PubChem CID 70671
IUPAC Name benzyl(triphenyl)phosphanium;chloride
SMILES [Cl-].C(C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Spezifikation

Melting Point ∼330°C (decomposition)
Flash Point >300°C (572°F)
Odor Odorless
Quantity 100 g
UN Number UN3464
Beilstein 3599868
Sensitivity Hygroscopic
Solubility Information Soluble in water and alcohol. Insoluble in acetone.
Formula Weight 388.88
Percent Purity 99%
Chemical Name or Material Benzyltriphenylphosphonium chloride
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.