missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Packaging | Bottle |
| Analyte or Component Names | Be |
| Concentration | 10±0.2 g/L |
| Type | Mass Spectrometry Standard |
| Element | Be |
| Solvent or Matrix | Nitric Acid/Hydrogen Fluoride |
| Quantity | 100 mL |
Safety and Handling
Product Identifier
- Single-element Standard Solution for ICP. Beryllium (Be) 10000mg/l in HNO3 5%,HF 1%
- Danger
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- Carcinogenicity Category 1B
- Specific target organ toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
- H290-May be corrosive to metals.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H350-May cause cancer.
- H372-Causes damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- P202-Do not handle until all safety precautions have been read and understood.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
- P390-Absorb spillage to prevent material damage.
- P501-Dispose of contents/container to .
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
