missing translation for 'onlineSavingsMsg'
Learn More

Beryllium Standard: Be @ 10 μg/mL in 2% HNO3, VHG™

Product Code. 17929349
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17929349 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17929349 Supplier VHG Labs Supplier No. VHGLBEN100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Beryllium Standard: Be @ 10 μg/mL in 2% HNO3
TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Beryllium
CAS 7440-41-7
Methods ICP/ICP-MS
Analyte or Component Names Beryllium
Shipping Condition Room Temperature
Molecular Formula Be
Concentration 10 μg/mL
Certifications/Compliance ISO 17034
Synonym Glucinium, Beryllium dust, Beryllium metal, Beryllium powder
InChI Formula InChI=1S/Be
SMILES [Be]
IUPAC Name beryllium
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 9.012
Quantity 100 mL
Formula Weight 9.012 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.