missing translation for 'onlineSavingsMsg'
Learn More

Beryllium Standard: Be @ 1000 μg/g in Hydrocarbon Oil, VHG™

Product Code. 30041170
Change view
Click to view available options
Quantity:
50 g
200 g
Unit Size:
200g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041170 50 g 50g
30040639 200 g 200g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30041170 Supplier VHG Labs Supplier No. VHGOBE100050G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones
Normativa: es posible que la compra de este producto esté sujeta a restricciones y necesitemos solicitarle documentación adicional.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Beryllium Standard: Be @ 1000 μg/g in Hydrocarbon Oil
TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material Beryllium
CAS 7440-41-7
Methods ICP/XRF/RDE
Analyte or Component Names Beryllium
Shipping Condition Room Temperature
Molecular Formula Be
Certifications/Compliance ISO 17034
Synonym Glucinium, Beryllium dust, Beryllium metal, Beryllium powder
InChI Formula InChI=1S/Be
SMILES [Be]
IUPAC Name beryllium
Solvent or Matrix 75 cSt Mineral Oil
Molecular Weight (g/mol) 9.012
Quantity 50 g
Formula Weight 9.012 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.