missing translation for 'onlineSavingsMsg'
Learn More

BES (White Crystalline Powder), Fisher BioReagents

Product Code. 11448063
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11448063 100 g Poly Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11448063 Supplier Fisher BioReagents Supplier No. BP501100
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • Used to make BES buffer with a useful pH range of 6.4-7.8 for biological applications
  • Literature describes BES buffer as preferred buffer for higher transfection efficiency of plasmid DNA
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10191-18-1
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
MDL Number MFCD00007533
InChI Key AJTVSSFTXWNIRG-UHFFFAOYSA-N
Synonym N, N-bis (2-Hydroxyethyl)-2-Amino-Ethanesulfonic Acid, N, N-bis (2-Hydroxyethyl) Taurine
PubChem CID 73243
ChEBI CHEBI:39045
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethane-1-sulfonic acid
SMILES OCCN(CCO)CCS(O)(=O)=O

Specifications

Melting Point 152°C
Color White
pH 2.5 to 5.0
Physical Form Solid
Quantity 100 g
DNase DNase free
Assay Percent Range ≥99 %
Loss on Drying 1% max.
Packaging Poly Bottle
Protease Protease free
Formula Weight 213.25
Percent Purity ≥99%
Chemical Name or Material BES
Show More Show Less
compliance-icons
Product Identifier
  • BES
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sikkerhedssætning
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.