missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta-1,3-N-Acetylglucosaminyltransferase 6/B3GNT6 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92200-0.1ml
This item is not returnable.
View return policy
Description
Beta-1,3-N-Acetylglucosaminyltransferase 6/B3GNT6 Polyclonal antibody specifically detects Beta-1,3-N-Acetylglucosaminyltransferase 6/B3GNT6 in Rat samples. It is validated for Western Blot
Specifications
| Beta-1,3-N-Acetylglucosaminyltransferase 6/B3GNT6 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| B3Gn-T6, beta-1,3-Gn-T6, Beta-1,3-N-acetylglucosaminyltransferase 6, beta-1,3-N-acetylglucosaminyltransferase protein, beta3Gn-T6, BGnT-6, Core 3 synthase, EC 2.4.1.-, IMAGE:4907098, MGC119334, MGC119336, MGC119337, UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6, UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6 (core 3 synthase) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 30-200 of human B3GNT6 (NP_619651.3). FLQAPRSPREERSPQEETPEGPTDAPAADEPPSELVPGPPCVANASANATADFEQLPARIQDFLRYRHCRHFPLLWDAPAKCAGGRGVFLLLAVKSAPEHYERRELIRRTWGQERSYGGRPVRRLFLLGTPGPEDEARAERLAELVALEAREHGDVLQWAFADTFLNLTLK | |
| 0.1 mL | |
| Endocrinology, Signal Transduction | |
| 192134 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction