missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-1,4-Galactosyltransferase 1/B4GalT1 Rabbit anti-Human, Clone: 1, Novus Biologicals™
Rabbit Monoclonal Antibody
367.00 € - 552.00 €
Specifications
| Antigen | beta-1,4-Galactosyltransferase 1/B4GalT1 |
|---|---|
| Clone | 1 |
| Dilution | ELISA 1:5000-1:10000, Sandwich ELISA Capture 1:250-1:2000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18277017
|
Novus Biologicals
NBP2-89848-50UL |
50 μg |
367.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18201376
|
Novus Biologicals
NBP2-89848-100UL |
100 μg |
552.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
beta-1,4-Galactosyltransferase 1/B4GalT1 Monoclonal specifically detects beta-1,4-Galactosyltransferase 1/B4GalT1 in Human samples. It is validated for ELISA, Sandwich ELISA Capture.Specifications
| beta-1,4-Galactosyltransferase 1/B4GalT1 | |
| ELISA 1:5000-1:10000, Sandwich ELISA Capture 1:250-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 1,4- galactosyltransferase, polypeptide 1, B4GAL-T1, beta-1,4-galactosyltransferase 1, Beta-1,4-GalTase 1, Beta4Gal-T1, betaGlcNAc beta, CDG2D, EC 2.4.1, GGTB2DKFZp686N19253, glycoprotein-4-beta-galactosyltransferase 2, GT1, GTB, lactose synthase, MGC50983, UDP-Gal, UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 1, UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 1 | |
| Recombinant Human beta-1,4-Galactosyltransferase 1/B4GalT1 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| 0.2 um filtered solution in PBS | |
| 2683 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title