missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta 2 Adaptin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99455-50ul
This item is not returnable.
View return policy
Description
Beta 2 Adaptin Polyclonal antibody specifically detects Beta 2 Adaptin in Human samples. It is validated for Western Blot
Specifications
| Beta 2 Adaptin | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Adapter-related protein complex 2 beta subunit, Adaptor protein complex AP-2 subunit beta, adaptor-related protein complex 2, beta 1 subunit, ADTB2adaptin, beta 2 (beta), AP105B, AP2-BETA, Beta-2-adaptin, Beta-adaptin, CLAPB1AP-2 complex subunit beta, Clathrin assembly protein complex 2 beta large chain, clathrin-associated/assembly/adaptor protein, large, beta 1, DKFZp781K0743, Plasma membrane adaptor HA2/AP2 adaptin beta subunit | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Beta 2 Adaptin. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 163 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction