Learn More
beta-Actin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 13 publications
Brand:  Novus Biologicals NB600-505
 View more versions of this product
Product Code. 18715373
  • 524.00 € / 0.10mg
Description
beta-Actin Polyclonal specifically detects beta-Actin in Human, Mouse, Rat, Amphibian samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Electron Microscopy.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
beta-Actin
Polyclonal
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1
Rabbit
42 kDa
0.1 mg
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Loading Controls, Signal Transduction, Stem Cell Markers
60
Human, Mouse, Rat, Amphibian, Bovine, Guinea Pig, Goat, Hamster, Sheep
IgG
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay
1 mg/ml
Western Blot 1:1000-1:4000, ELISA 1:10000-1:40000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:500-1:2000, Immunohistochemistry-Paraffin 1:50 - 1:200, Electron Microscopy
P60709
ACTB
beta-Actin Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to C-Terminal region near amino acids 350-375 of Human beta Actin. (Uniprot: P60709)
Affinity Purified
RUO
Primary
Expected to cross-react with a wide range of species including most vertebrates and some yeast due to sequence identity.
Store at -20°C. Avoid freeze-thaw cycles.