Learn More
beta Amyloid Antibody (CNTO 2125) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-28180JF669
Product Code. 30163852
  • 419.00 € / 0.10mL
Description
beta Amyloid Monoclonal antibody specifically detects beta Amyloid in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
beta Amyloid
Monoclonal
Janelia Fluor 669
Abeta, Amyloid beta, beta-amyloid peptide
Amyloid Beta
0.1 mL
Alzheimers Research, Apoptosis, Cardiovascular Biology, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific, Signal Transduction
351
Store at 4°C in the dark.
IgG1
Flow Cytometry, ELISA, Functional Assay
CNTO 2125
50mM Sodium Borate
Human
Protein A purified
RUO
Primary
Human
Purified