missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Amyloid Mouse anti-Human, Porcine, Clone: 5C2A1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60342-1-IG-150UL
This item is not returnable.
View return policy
Description
Amyloid beta peptide (Abeta) is the major constituent of amyloid plaques in the brains of individuals afflicted with Alzheimer′s disease. This peptide is generated from the beta-amyloid precursor protein (beta APP) in a two-step process. The first step involves cleavage of the extracellular, amino-terminal domain of beta APP. Protein cleavage is performed by an aspartyl protease termed beta-secretase (BACE). This enzyme is synthesized as a propeptide that must be modified to the mature and active form by the prohormone convertase, furin. Beta APP cleavage by the mature form of BACE results in the cellular secretion of a segment of beta APP and a membrane-bound remt. This remt is then processed by another protease termed gamma-secretase. Gamma-secretase cleaves an intra-membrane site in the carboxyl-terminal domain of beta APP, thus generating the amyloid beta peptide. Gamma-secretase is believed to be a multi-subunit complex containing presenilin-1 and 2 as central components. Found associated with the presenilins is the transmembrane glycoprotein nicastrin. Nicastrin has been found to bind to the carboxyl-terminus of betaAPP and helps to modulate the production of the amyloid beta peptide Also found in the neurofibrillary lesions in Alzheimer′s disease is the protein termed Tau. Tau is a neuronal microtubule-associated protein found predomitly on axons. The function of tau is to promote tubulin polymerization and stabilize microtubules. Tau, in its hyperphosphorylated form, is the major component of paired helical filaments (PHF), which is the building block of neurofibrillary lesions in Alzheimer′s disease brain.Specifications
| beta Amyloid | |
| Monoclonal | |
| 1.12 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P05067, P79307 | |
| APP | |
| beta Amyloid Fusion Protein Ag0769 | |
| 150 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| 5C2A1 | |
| Unconjugated | |
| APP | |
| A4, AAA, ABETA, ABPP, AD1, AICD 50, AICD 57, AICD 59, AID(50), AID(57), AID(59), Amyloid beta A4 protein, amyloid P, APP, APPI, beta Amyloid, CTFgamma, CVAP, Gamma CTF(50) C31, PN II, PN2, PreA4, Protease nexin II, S APP alpha Soluble APP beta | |
| Mouse | |
| Protein A | |
| RUO | |
| 351, 397663 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction