missing translation for 'onlineSavingsMsg'
Learn More

beta-Catenin Antibody (9F2), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-53257-100UG

 View more versions of this product

Product Code. 18129539

  • 508.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

beta-Catenin Monoclonal specifically detects beta-Catenin in Human, Bovine, Canine, Chicken samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

beta-Catenin
Monoclonal
0.2 mg/mL
Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, SDS-Page
beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923
Mouse
92 kDa
100 μg
Cancer, Cellular Markers, Extracellular Matrix, Mucosal Immunology, Neuroscience, Phospho Specific, Stem Cell Signaling Pathway, Wnt Signaling Pathway
1499
Store at 4C.
IgG1 κ
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page
9F2
Unconjugated
10mM PBS with 0.05% BSA with 0.05% Sodium Azide
CTNNB1
Recombinant chicken Beta-catenin
Protein A or G purified
RUO
Primary
Human, Bovine, Canine, Chicken
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only