missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta Catenin Mouse, Clone: 12F7, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 478.00 €
Specifications
| Antigen | Beta Catenin |
|---|---|
| Clone | 12F7 |
| Concentration | 200μg/mL |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15949857
|
EnquireBio
1499-MSM2-P0 |
20 μg | |||||||
|
15959857
|
EnquireBio
1499-MSM2-P1 |
100 μg |
554.40 € 478.00 € / 100µg Save 76.40 € 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| Beta Catenin | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| 1499 | |
| Recombinant fusion protein consisting of the N-terminal half of human beta-Catenin fused to maltose binding protein | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 92kDa |
| 12F7 | |
| Immunofluorescence | |
| Unconjugated | |
| RUO | |
| NP_001895 | |
| CTNNB1 | |
| IgG1 κ | |
| Protein A/G Purified | |
| Cell surface and cytoplasmic | |
| HeLa or MCF-7 cells. Breast carcinoma |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title