missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Galactosidase, Armenian Hamster anti-E. coli, Clone: 13R4, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
250.00 € - 655.00 €
Specifications
| Antigen | beta Galactosidase |
|---|---|
| Clone | 13R4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, Flow Cytometry, Control |
| Applications | Western Blot, Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18317100
|
Novus Biologicals
NBP2-81033-0.025MG |
0.025 mg |
250.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18205719
|
Novus Biologicals
NBP2-81033-0.2MG |
0.2 mg |
655.00 €
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
beta Galactosidase Monoclonal specifically detects beta Galactosidase in E. coli samples. It is validated for Western Blot, Flow Cytometry, Control.Specifications
| beta Galactosidase | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Armenian Hamster | |
| E. coli | |
| EC 3.2.1.23; b-gal | |
| IgG λ | |
| Protein A purified | |
| 13R4 recognizes wild-type E. coli beta-galactosidase. |
| 13R4 | |
| Western Blot, Flow Cytometry, Control | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| beta Galactosidase. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title