missing translation for 'onlineSavingsMsg'
Learn More

(+)-Beta-Methyl-Alpha,Alpha-diphenyl-4-morpholinebutanoic Acid, TRC

Product Code. 30477768
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477768 5 mg 5mg
30474785 10 mg 10mg
30466733 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30477768 Supplier Toronto Research Chemicals Supplier No. TRCM3030935MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (+)-beta-Methyl-alpha,alpha-diphenyl-4-morpholinebutanoic Acid
Molecular Formula C21H25NO3
Quantity 5 mg
Synonym (S)-3-Methyl-4-morpholino-2,2-diphenylbutanoic Acid
InChI Formula InChI=1S/C21H25NO3/c1-17(16-22-12-14-25-15-13-22)21(20(23)24,18-8-4-2-5-9-18)19-10-6-3-7-11-19/h2-11,17H,12-16H2,1H3,(H,23,24)/t17-/m1/s1
SMILES C[C@@H](C(C1=CC=CC=C1)(C2=CC=CC=C2)C(O)=O)CN3CCOCC3
IUPAC Name (S)-3-methyl-4-morpholino-2,2-diphenylbutanoic acid
Molecular Weight (g/mol) 339.43
Formula Weight 339.18
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.