missing translation for 'onlineSavingsMsg'
Learn More

Beta-Methylphenethylamine Hydrochloride, TRC

Product Code. 30468719
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30468719 10 g 10g
30468429 25 g 25g
30468027 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30468719 Supplier Toronto Research Chemicals Supplier No. TRCM32478010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20388-87-8
Molecular Formula C9 H13 N . Cl H
Molecular Weight (g/mol) 171.67
Synonym β-Methylphenethylamine Hydrochloride, Benzeneethanamine, β-methyl-, hydrochloride (1:1), Benzeneethanamine, β-methyl-, hydrochloride (9CI), Phenethylamine, β-methyl-, hydrochloride (8CI), 2-Phenyl-1-propanamine hydrochloride, 2-Phenyl-1-propanamine monohydrochloride, 2-Phenylpropylamine hydrochloride, β-Methylbenzeneethanamine hydrochloride
IUPAC Name 2-phenylpropan-1-amine;hydrochloride
SMILES Cl.CC(CN)c1ccccc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C9H13N.ClH/c1-8(7-10)9-5-3-2-4-6-9;/h2-6,8H,7,10H2,1H3;1H
Formula Weight 171.0815
Chemical Name or Material R-beta-Methylphenylethylamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.