missing translation for 'onlineSavingsMsg'
Learn More

Beta-Pinene 1000 μg/mL in Isopropanol, Dr. Ehrenstorfer

Product Code. 30309488
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30309488 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309488 Supplier Dr. Ehrenstorfer Supplier No. DREGS09010045IP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material beta-Pinene
CAS 127-91-3
Analyte or Component Names beta-Pinene
Shipping Condition Room Temperature
Molecular Formula C10 H16
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym 2(10)-Pinene (8CI), (±)-2(10)-Pinene, (±)-6,6-Dimethyl-2-methylenebicyclo[3.1.1]heptane, (±)-β-Pinene, 6,6-Dimethyl-2-methylenebicyclo[3.1.1]heptane, NSC 21447, NSC 406265, NSC 59190, Nopinen, Nopinene, PC 600, PC 600 (pesticide), Pseudopinen, Pseudopinene, Terebenthene, beta-Pinene, β-Pinene
InChI Formula InChI=1S/C10H16/c1-7-4-5-8-6-9(7)10(8,2)3/h8-9H,1,4-6H2,2-3H3
SMILES CC1(C)C2CCC(=C)C1C2
IUPAC Name 6,6-dimethyl-4-methylidenebicyclo[3.1.1]heptane
Solvent or Matrix Isopropanol
Molecular Weight (g/mol) 136.23
Quantity 5 x 1 mL
Formula Weight 136.1252 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.