missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BetaBasic™ 18 Guard Cartridges

Product Code. 11592841 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric):
2.1 mm
3 mm
4 mm
Length (Metric):
10 mm
Particle Size:
3 μm
5 μm
Unit Size:
Pack of 4
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Length (Metric) Particle Size Unit Size
11592841 4 mm 10 mm 5 μm Pack of 4
10197984 2.1 mm 10 mm 3 μm Pack of 4
11572841 3 mm 10 mm 3 μm Pack of 4
11582841 4 mm 10 mm 3 μm Pack of 4
10419634 2.1 mm 10 mm 5 μm Pack of 4
10438094 3 mm 10 mm 5 μm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11592841 Supplier Thermo Scientific™ Supplier No. 71505014001
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Available in limited dimensions. Please contact Customer Service for more details.

Thermo Scientific™ BetaBasic™ 18 Guard Cartridges are available in limited dimensions. Please contact Customer Service for more details.

  • The 10mm design offers maximum protection with minimal increase in retention
  • Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
  • Highly efficient and reproducible Thermo Scientific columns
  • 150Å pore size for small molecules, peptides and protein digests
  • Superb pH stability

Requires:

Thermo Scientific Unigard™ direct connection or stand-alone holder, sold separately.

TRUSTED_SUSTAINABILITY

Specifications

Carbon Load 13%
pH 2 to 12
Particle Size 5 μm
Length (Metric) 10 mm
Pore Size 150Å
Surface Area 200 m2/g
Diameter (Metric) 4 mm
For Use With (Equipment) Uniguard Direct-connection and Stand Alone Holder
Endcapped Yes
USP Type L1
Product Line Beta
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.