missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BetaBasic™ 18 Javelin™ Guard Columns

Product Code. 11508730 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric) Inner:
2.1 mm
4 mm
Length (Metric):
10 mm
20 mm
Particle Size:
5 μm
Unit Size:
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Length (Metric) Particle Size unitSize
11508730 4 mm 10 mm 5 μm Pack of 4
11588720 2.1 mm 10 mm 5 μm Pack of 4
11563251 2.1 mm 20 mm 5 μm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11508730 Supplier Thermo Scientific™ Supplier No. 71505014006
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Offer convenient, easy to use protection for analytical HPLC columns

  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
  • Highly efficient and reproducible Thermo Scientific columns
  • 150Å pore size for small molecules, peptides and protein digests
  • Superb pH stability

Order Info

Javelin guards are available in other Thermo Scientific phases. Please call your Customer Service Representative for more information.

TRUSTED_SUSTAINABILITY

Specifications

Particle Size 5 μm
Length (Metric) 10 mm
Diameter (Metric) Inner 4 mm
Stationary Phase BetaBasic 18
Product Line Beta
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.