missing translation for 'onlineSavingsMsg'
Learn More

Betaine, TRC

Product Code. 30007764
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007764 100 g 100g
30012053 50 g 50g
17957934 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007764 Supplier Toronto Research Chemicals Supplier No. TRCB325005100G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-43-7
Molecular Formula C5 H11 N O2
Molecular Weight (g/mol) 117.15
Synonym Methanaminium, 1-carboxy-N,N,N-trimethyl-, inner salt, Ammonium compounds, substituted, (carboxymethyl)trimethyl-, hydroxide, inner salt (7CI), Betaine (8CI), Methanaminium, 1-carboxy-N,N,N-trimethyl-, hydroxide, inner salt, (Carboxymethyl)trimethylammonium hydroxide inner salt, (Trimethylammonio)acetate, Abromine, Aminocoat, Aquadew AN 100, Auqadew AN 100, Betafin, Betafin BCR, Betafin BP, Betafin BP 20, Bluestim, Cystadane, Fencaijian, FinnStim, Glycine betaine, Glycine, trimethylbetaine, Glycocoll betaine, Glycylbetaine, Greenstim, Loramine AMB 13, Lycine, N,N,N-Trimethylglycine, Oxyneurine, Rubrine C, Trimethylbetaine, Trimethylglycine, Trimethylglycocoll, α-Earleine
IUPAC Name 2-(trimethylazaniumyl)acetate
SMILES C[N+](C)(C)CC(=O)[O-]

Specifications

Analyte or Component Names Betaine (Trimethylglycine)
Shipping Condition Room Temperature
InChI Formula InChI=1S/C5H11NO2/c1-6(2,3)4-5(7)8/h4H2,1-3H3
Solution Type Neat
Industry Type Pharmaceutical
Quantity 100 g
Formula Weight 117.079
Chemical Name or Material Betaine (Trimethylglycine)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.