missing translation for 'onlineSavingsMsg'
Learn More

Betaxolol hydrochloride, MedChemExpress

Product Code. 16454347
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16454347 50 mg 50mg
16428468 10 mg 10mg
16428276 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16454347 Supplier Medchem Express Supplier No. HYB0381A
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Betaxolol Hydrochloride is a selective beta1 adrenergic receptor blocker that can be used for the research of hypertension and glaucoma.

Betaxolol Hydrochloride is a selective beta1 adrenergic receptor blocker that can be used for the research of hypertension and glaucoma.

Specifications

Chemical Name or Material Betaxolol hydrochloride
CAS 63659-19-8
Color White
Purity Grade Notes Research
Molecular Formula C18H30ClNO3
Quantity 50 mg
Synonym SL75212
Solubility Information DMSO : ≥ 100 mg/mL (290.79 mM) ∣H2O : 10 mg/mL (29.08 mM; Need ultrasonic)
SMILES OC(CNC(C)C)COC1=CC=C(CCOCC2CC2)C=C1.[H]Cl
Molecular Weight (g/mol) 343.89
Formula Weight 343.89
Percent Purity 98.69%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.