missing translation for 'onlineSavingsMsg'
Learn More

Betaxolol hydrochloride, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18722724
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18722724 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18722724 Supplier Tocris Bioscience Supplier No. 0906/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selective β1 antagonist

Selective β1-adrenoceptor antagonist.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63659-19-8
Molecular Formula C18H30ClNO3
Molecular Weight (g/mol) 343.892
InChI Key CHDPSNLJFOQTRK-UHFFFAOYSA-N
Synonym betaxolol hydrochloride, betaxolol hcl, betoptima, kerlone, betoptic, kerlon, lokren, betaxolo hydrochloride, betoptic s
PubChem CID 107952
ChEBI CHEBI:643228
IUPAC Name 1-[4-[2-(cyclopropylmethoxy)ethyl]phenoxy]-3-(propan-2-ylamino)propan-2-ol;hydrochloride
SMILES CC(C)NCC(COC1=CC=C(C=C1)CCOCC2CC2)O.Cl

Specifications

Quantity 50 mg
Formula Weight 343.89
Percent Purity >99%
Chemical Name or Material Betaxolol hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.