missing translation for 'onlineSavingsMsg'
Learn More

Betaxolol, MedChemExpress

Product Code. 16444347
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16444347 50 mg 50mg
16418468 10 mg 10mg
16418276 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16444347 Supplier Medchem Express Supplier No. HYB0381
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Betaxolol is a selective beta1 adrenergic receptor blocker that can be used for the research of hypertension and glaucoma.

Betaxolol is a selective beta1 adrenergic receptor blocker that can be used for the research of hypertension and glaucoma.

Specifications

Chemical Name or Material Betaxolol
CAS 63659-18-7
Color White
Purity Grade Notes Research
Molecular Formula C18H29NO3
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (325.28 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES OC(CNC(C)C)COC1=CC=C(CCOCC2CC2)C=C1
Molecular Weight (g/mol) 307.43
Formula Weight 307.43
Percent Purity 95.06%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.