missing translation for 'onlineSavingsMsg'
Learn More

Betaxolol, TRC

Product Code. 17972335
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17972335 10 mg 10mg
17901255 5 mg 5mg
17962215 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17972335 Supplier Toronto Research Chemicals Supplier No. TRCB32800010MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63659-18-7
Molecular Formula C18 H29 N O3
Molecular Weight (g/mol) 307.43
Synonym (±)-Betaxolol, 1-Isopropylamino-3-[4-(2-cyclopropylmethoxy-ethyl)phenoxy]-2-propanol, Betaxolol, Betoptic S, 2-Propanol, 1-[4-[2-(cyclopropylmethoxy)ethyl]phenoxy]-3-[(1-methylethyl)amino]-
IUPAC Name 1-[4-[2-(cyclopropylmethoxy)ethyl]phenoxy]-3-(propan-2-ylamino)propan-2-ol
SMILES CC(C)NCC(O)COc1ccc(CCOCC2CC2)cc1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H29NO3/c1-14(2)19-11-17(20)13-22-18-7-5-15(6-8-18)9-10-21-12-16-3-4-16/h5-8,14,16-17,19-20H,3-4,9-13H2,1-2H3
Formula Weight 307.2147
Percent Purity >95%
Chemical Name or Material Betaxolol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.