missing translation for 'onlineSavingsMsg'
Learn More

BF738735, MedChemExpress

Product Code. p-200096186
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16492628

Brand: Medchem Express HYU00426

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BF738735 is a phosphatidylinositol 4-kinase III beta (PI4KIIIβ) inhibitor with an IC50 of 5.7 nM.

BF738735 is a phosphatidylinositol 4-kinase III beta (PI4KIIIβ) inhibitor with an IC50 of 5.7 nM.

Specifications

Chemical Name or Material BF738735
CAS 1436383-95-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H19FN4O3S
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (293.11 mM; Need ultrasonic)
SMILES OC1=CC=C(C2=C(C)N=C3C(NCC4=CC=CC(S(=O)(C)=O)=C4)=NC=CN32)C=C1F
Molecular Weight (g/mol) 426.46
Formula Weight 426.46
Percent Purity 99.15%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.