missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BFAR Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92687-0.02ml
This item is not returnable.
View return policy
Description
BFAR Polyclonal antibody specifically detects BFAR in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| BFAR | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:100 | |
| BARRNF47RING finger protein 47, bifunctional apoptosis inhibitor, bifunctional apoptosis regulator | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human BFAR (NP_057645.1). MEEPQKSYVNTMDLERDEPLKSTGPQISVSEFSCHCCYDILVNPTTLNCGHSFCRHCLALWWASSKKTECPECREKWEGFPKVSILLRDAIEKLFPDAIRLRFEDIQQNNDIVQSLAAFQKYGNDQIPLAPNTGRANQQM | |
| 0.02 mL | |
| Apoptosis, Zinc Finger | |
| 51283 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction