missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BHLHB5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-39039-25ul
This item is not returnable.
View return policy
Description
BHLHB5 Polyclonal specifically detects BHLHB5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| BHLHB5 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
| Q8NFJ8 | |
| BHLHE22 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: EDDLFLHKSLSASTSKRLEAAFRSTPPGMDLSLAPPPRERPASSSSSPLGCFEPADPEGAGLLLPPP | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Basic Helix-Loop-Helix Domain Containing, Class B, 5, basic helix-loop-helix family, member e22, Beta3, BHLHE22, CAGL85, Class B Basic Helix-Loop-Helix Protein 5, Class E Basic Helix-Loop-Helix Protein 22, TNRC20, Trinucleotide Repeat Containing 20, Trinucleotide Repeat-Containing Gene 20 Protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 27319 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction