missing translation for 'onlineSavingsMsg'
Learn More

BHLHE41 Monoclonal Antibody (OTI10B11), TrueMAB™, OriGene

Product Code. 17536213
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17536213 30 μL 30µL
17516213 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17536213 Supplier Origene Technologies Supplier No. TA806146S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BHLHE41 Monoclonal antibody specifically detects BHLHE41 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen BHLHE41
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI10B11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene FSIP1
Gene Accession No. Q8NA03
Gene Alias 2-Dec; 6430520M22Rik; basic helix-loop-helix domain containing, class B, 3; basic helix-loop-helix domain containing, class B3; basic helix-loop-helix family member e41; basic helix-loop-helix family, member e41; bHLH transcriptional factor Dec2; Bhlhb2l; Bhlhb3; bHLHe41; Class B basic helix-loop-helix protein 3; class E basic helix-loop-helix protein 41; Dec2; differentially expressed in chondrocytes protein 2; Enhancer-of-split and hairy-related protein 1; hDEC2; mDEC2; SHARP1; SHARP-1
Host Species Mouse
Immunogen Full length human recombinant protein of human FSIP1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161835
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.