missing translation for 'onlineSavingsMsg'
Learn More

BICINE, 0.5M buffer soln., pH 9.0, Thermo Scientific™

Product Code. 15425149
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
Product Code. Quantity unitSize
15425149 250 mL 250mL
15415149 100 mL 100mL
2 options
This item is not returnable. View return policy

Product Code. 15425149

Brand: Thermo Scientific Alfa Aesar J62838.AK

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H12NNaO4
Molecular Weight (g/mol) 185.16
MDL Number MFCD00004295
InChI Key MFBDBXAVPLFMNJ-UHFFFAOYSA-M
PubChem CID 8761
ChEBI CHEBI:40957
IUPAC Name 2-[bis(2-hydroxyethyl)amino]acetic acid
SMILES [Na+].OCCN(CCO)CC([O-])=O

Specifications

Concentration 0.5 M
pH 9.0
Physical Form Liquid
Quantity 250 mL
Chemical Name or Material BICINE buffer soln.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.