missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Bicine
Description
Zwitterionic buffer used in a variety of molecular biology applications such as protein resolution, protein crystallization and SDS-PAGE.
Zwitterionic buffer used in a variety of molecular biology applications such as protein resolution, protein crystallization and SDS-PAGE.
pKa 25°C: 8.3
pH Range: 7.6-9.0
Melting Range: 190-192°C
Specifications
Specifications
| Chemical Name or Material | Bicine |
| CAS | 150-25-4 |
| Quantity | 100 g |
| Molecular Formula | C6H13NO4 |
| Grade | Ultra Pure |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?